Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADO Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS2640R

Catalog No. 50-198-4865


Add to cart

Description

Description

Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.
Specifications

Specifications

ADO
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ADO
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
84890
ADO
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q96SZ5
2-aminoethanethiol (cysteamine) dioxygenase; 2-aminoethanethiol dioxygenase; ADO; C10orf22; cysteamine (2-aminoethanethiol) dioxygenase (ADO); Cysteamine dioxygenase; Gm237
KLH conjugated synthetic peptide derived from human ADO .
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit