Learn More
Invitrogen™ AdoHcyase 2 Monoclonal Antibody (2E7D9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549301
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human MCF-7 whole cell, rat brain tissue, rat pancreas tissue, mouse brain tissue, mouse pancreas tissue. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene interacts with inositol 1,4,5-trisphosphate receptor, type 1 and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding two different isoforms have been found for this gene.
Specifications
AdoHcyase 2 | |
Monoclonal | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
B5DFN2, O43865, Q80SW1 | |
AHCYL1 | |
E.coli-derived human IRBIT/AHCYL1 recombinant protein (Position: E14-K57). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
2E7D9 | |
Unconjugated | |
AHCYL1 | |
1110034F20Rik; AA409031; AA414901; adenosylhomocysteinase 2; adenosylhomocysteinase 2; S-adenosylhomocysteine hydrolase-like protein 1; adenosylhomocysteinase like 1; adenosylhomocysteinase-like 1; adoHcyase 2; Ahcyl1; Ahcyl1-v1; Ahcyl1-v2; Ahcy-rs3; cb586; DCAL; DC-expressed AHCY-like molecule; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IP3R binding protein released with inositol 1,4,5-trisphosphate; IP3R-binding protein released with inositol 1,4,5-trisphosphate; Irbit; PPP1R78; PRO0233; protein phosphatase 1, regulatory subunit 78; putative adenosylhomocysteinase 2; s adenosylhomocysteine hydrolase; S-adenosyl homocysteine hydrolase homolog; S-adenosylhomocysteine hydrolase, related sequence 3; S-adenosylhomocysteine hydrolase-like 1; S-adenosylhomocysteine hydrolase-like protein 1; S-adenosylhomocysteine hydrolase-like protein variant 1; S-adenosylhomocysteine hydrolase-like protein variant 2; S-adenosyl-L-homocysteine hydrolase 2; wu:fj66f02; XPVKONA | |
Mouse | |
Antigen Affinity Chromatography | |
RUO | |
10768, 229709, 362013 | |
-20°C | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.