Learn More
ADORA2A 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Supplier:  Abnova Corporation H00000135T01
Catalog No. 89-000-213
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
ADORA2A 293T Cell Transient Overexpression Lysate(Denatured)
45.43
pCMV-ADORA2A full-length
100 μL
Human
293T
NM_000675
6.8
Transient overexpression cell Lysate was tested with Anti-ADORA2A antibody (H00000135-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'