Learn More
BioVision IncSupplier Diversity Partner PicoProbe™ ADP Assay Kit (Fluorometric): PicoProbe™ ADP Assay Kit (Fluorometric): Fast, sensitive, HTP compatible, fluorometric ADP assay with detection limit as low as 10 pmol or 0.05 µM of ADP. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K211100
PicoProbe™ ADP Assay Kit (Fluorometric): Adenosine diphosphate (ADP) is an organic molecule that plays a critical role in cellular metabolism and energy transfer reactions. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. BioVision's PicoProbe™ ADP Assay Kit provides a sensitive fluorescence method to detect ADP. The kit is suitable for measuring ADP levels in various samples, including the ones that contain reducing substances, which may interfere with oxidase-based assays. In this assay, ADP, in the presence of ADP Enzyme Mix is converted to an intermediate, which reacts with the fluorescent probe to generate a strong stable signal at Ex/Em = 535/587 nm. BioVision's PicoProbe™ ADP Assay kit provides an ultrasensitive, fast and high-throughput ready assay for detection of as low as 10 pmol or 0.05 µM of ADP in samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9622