Learn More
BioVision IncSupplier Diversity Partner ADP Colorimetric Assay Kit II: ADP Colorimetric Assay Kit II : Measure ADP concentration in animal tissues & cells within 30 min. Simple, Convenient, Sensitive & High-throughput. Detection limit: less than 20 µM. 100 wells. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K356100
ADP Colorimetric Assay Kit II: ADP is a product of ATP dephosphorylation and it can be rephosphorylated to ATP. Dephosphorylation and rephosphorylation occur via various phosphorylases and kinases. ADP is stored in platelets and can be released to interact with variety of purinergic receptors. ADP levels regulate several enzymes involved in intermediary metabolism. The luciferase system is unstable and luminescence equipment is not generally available in most laboratories. BioVision ADP Assay kit II is suitable for measuring ADP levels in samples that contain reducing substances, which may interfere with oxidase-based assays. In this assay, ADP in the presence of ADP Enzyme Mix is converted to an intermediate, which reduces a colorless Probe to a colored product with strong absorbance at 450 nm. ADP Assay Kit II is simple, fast and high-throughput ready. It can detect less than 20 ?M of ADP in samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9715