Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADPRH Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549268
Description
Predicted to react with bovine based on sequence homology.
Catalyzes the reverse reaction of mono-ADP-ribosylation.Specifications
ADPRH | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P54922 | |
ADPRH | |
KLH conjugated synthetic peptide between 306-334 amino acids from the C-terminal region of human ADPRH. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ADPRH | |
[Protein ADP-ribosylarginine] hydrolase, ADP-ribosylarginine hydrolase, ADP-ribose-L-arginine cleaving enzyme, ADPRH, ARH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
141 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction