Learn More
ADRA1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 224191AP
Catalog No. 50-173-3863
Add to cart
Description
Description
The alpha-1b adrenoceptor is an Adrenergic Receptor that causes contraction of smooth muscle cells and thereby controls vascular tone, blood pressure, and accelerates the development of atherosclerosis. Decreased alpha-1b adrenoceptor is associated with benign prostatic hyperplasia (BPH). This adrenergic receptor also acts as a proto-oncogene when transfected into cell lines where constitutive activation of the receptor induces neoplastic transformation. The alpha-1b adrenoceptor has been documented in adrenal, aorta, blood, various brain regions, eye, heart, kidney, liver, prostate, spinal cord, and spleen. ESTs have been isolated from kidney (normal and cancer) and lung carcinoma libraries.
Specifications
Specifications
ADRA1B
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15823, P35368, P97717
ADRA1, ADRA1B, Alpha 1B adrenergic receptor, Alpha 1B adrenoceptor, Alpha 1B adrenoreceptor, ALPHA1BAR
Rabbit
IgG
150 μL
-20°C
Primary
11548, 147, 24173
Flow Cytometry, Western Blot
Unconjugated
ADRA1B
Liquid
Adra1b
ADRA1B Fusion Protein Ag18138
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat