Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRBK1 (GRK2) Recombinant Human Protein Non-distribution product as customer accommodation.

ADRBK1 (GRK2) Recombinant Human Protein

$432.00

Specifications

Accession Number NP_001610.2
Common Name ADRBK1 (GRK2) Recombinant Human Protein
Expression System Baculovirus
Gene Alias ADRBK1 (GRK2)
Molecular Weight 82.3kDa
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3361 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3361
10μg Each for $432.00
Add to cart
 
Description

Description

ADRBK1 (GRK2) is a serine⁄threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs)
Specifications

Specifications

NP_001610.2
Baculovirus
82.3kDa
1 Each
95% by SDS-PAGE
–80°C
Human
ADRBK1 (GRK2) Recombinant Human Protein
ADRBK1 (GRK2)
ADRBK1 (GRK2) Recombinant Human Protein
ADRBK1 (GRK2) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit