Learn More
Alfa Aesar™ Adrenomedullin (16-31), Human, Pig
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66935MCR
 View more versions of this product
Catalog No. AAJ66935MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1865.18
Human, Pig
Solid
RUO
Adrenomedullin (16-31), human, pig
HPLC
1mg
Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-NH2 (Disulfide bridge Cys16-Cys21)
-30° to -10°C