Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adrenomedullin/ADM Mouse anti-Human, Clone: OTI8A1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270100
Description
Adrenomedullin/ADM Monoclonal antibody specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry.Specifications
Adrenomedullin/ADM | |
OTI8A1 | |
Unconjugated | |
Affinity Purified,Lyophilized | |
adrenomedullin, AMpreproadrenomedullin | |
ADM | |
Human recombinant protein fragment corresponding to amino acids 22-146 of human ADM(NP_001115) produced in E.coli. | |
20.2 kDa | |
Primary | |
100 μg | |
GPCR | |
Human |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 1:250 | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
133 | |
Mouse | |
IgG2b | |
Monoclonal | |
Immunogen affinity purified | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title