Learn More
CPC Scientific H-Tyr-Arg-Gln-Ser-Met-Asn-Asn-Phe-Gln-Gly-Leu-Arg-Ser-Phe-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ADRM001A
Encompass
Participating hormone in blood pressure control that also elicits a potent and long lasting hypotensive effect.SEQUENCE: H-Tyr-Arg-Gln-Ser-Met-Asn-Asn-Phe-Gln-Gly-Leu-Arg-Ser-Phe-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2 (trifluoroacetate salt)(Cys16 and 21 bridge)ONE-LETTER SEQUENCE: YRQSMNNFQGLRSFGCRFGTCTVQKLAHQIYQFTDKDKDNVAPRSKISPQGY-NH2 (Cys16 and 21 bridge)MOLECULAR FORMULA: C264H406N80O77S3MOLECULAR WEIGHT: 6028.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [148498-78-6]SYNONYMS: RESEARCH AREA: CardiovascularREFERENCES:K. Kitamura et al., BBRC, 192, 533 (1993)C. Nuki et al., BBRC, 196, 245 (1993)SKU(s): ADRM-001A, ADRM-001B
Catalog No. 50-210-8619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.