Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenomedullin Mouse anti-Human, Clone: HTA 91a/G2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524772

Catalog No. PIMA524772


Add to cart

Description

Description

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintece of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
Specifications

Specifications

Adrenomedullin
Monoclonal
1 mg/mL
PBS with 0.02% sodium azide; pH 7.4
P35318
adrenomedullin, AMpreproadrenomedullin
Mouse
IgM
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
133
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
HTA 91a/G2
Unconjugated
ADM
Liquid
ADM
Chemically synthesized full length adrenomedullin.
IgM
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit