Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenosterone, TCI America™

Click to view available options
1G
382-45-6
C19H24O3
300.398
MFCD00003606
RZRPTBIGEANTGU-IRIMSJTPSA-N
4-Androstene-3,11,17-trione, Reichstein′Cs Substance G
223997
CHEBI:2495
(8S,9S,10R,13S,14S)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11,17-trione
CC12CCC(=O)C=C1CCC3C2C(=O)CC4(C3CCC4=O)C
382-45-6
C19H24O3
300.398
MFCD00003606
RZRPTBIGEANTGU-IRIMSJTPSA-N
4-Androstene-3,11,17-trione, Reichstein′Cs Substance G
223997
CHEBI:2495
(8S,9S,10R,13S,14S)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11,17-trione
CC12CCC(=O)C=C1CCC3C2C(=O)CC4(C3CCC4=O)C
Quantity 1g
Melting Point 222°C
Color White-Yellow
Formula Weight 300.40
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Adrenosterone