Sino Biological™ ADSL Recombinant Human Protein, N-His tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human ADSL (Met 1-Leu 484) fused to polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 11287H07E5

 View more versions of this product

Catalog No. 11-287-H07E5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 57kd
    Purity ≥90% by SDS-PAGE
    Source Human
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Adenylosuccinate Lyase
    Gene Symbol ADSL
    Validated Application Assay Standard
    Protein Family Other proteins
    Product Type ADSL Recombinant Human Protein
    Format Lyophilized
    Gene ID (Entrez) 158
    Quantity 5μg
    Protein Tag His Tag
    Expression System E.coli

    Expressed from DNA sequence encoding human ADSL (Met 1-Leu 484) fused to polyhistidine tag at N-terminus

    • Formulation: PBS, 10% Glycerol, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit