Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Delicate, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21772
Econo Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Delicate, Stainless Steel. Econo Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with delicate, serrated tips and a length of 4-3/4 inches. They are sold in boxes of 12.
Catalog No. 50-143-7682