Learn More
Analytical Sales & Services Advantage Unassembled Maximum Recovery Vial Kit  DFS Item
Manufacturer:  Analytical Sales & Services 878789
Advantage Unassembled Maximum Recovery Vial Kit Contains: 9mm, 12 x 32, 1.2mL Clear Glass Tear Drop Screw Vials and 9mm Screw Caps with PTFE/Silicone/PTFE Liners. < 2µL Dead Volume
Catalog No. 50-646-332