Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advance Tabco™ Drop-In Sinks

Supplier:  Advance Tabco™ DI22012

 View more versions of this product

Catalog No. 19-999-284


Add to Cart

Includes: All models include 1.5 in. IPS basket drain and aluminum/stainless mounting hardware to secure sink to countertop.
Model DI-1-10 includes deck mounted 8.5 in. gooseneck faucet, 4 in. O.C.
Model DI-1-2812 includes deck mounted 12 in. swing spout faucet, 4 in. O.C.
Model DI-2-2012 includes deck mounted 8 in. swing spout faucet, 4 in. O.C.

Description

Description

Heavy-duty stainless-steel construction with deep-drawn seamless bowl design for efficient drainage

  • Feature deep-drawn, one-piece sink bowl for most efficient drainage
  • Heavy-gauge stainless steel polished to a satin finish on the bowl interior
  • Sound deadened bowls
  • Faucet supply is 0.5 in. IPS male thread

Order Info

Model DI-1-10 Bowl depth: 10 in. (25cm); Sink L x W: 10 x 14 in. (25 x 36cm) Model DI-2-2012 Bowl depth: 12 in. (30.5cm); Sink L x W: 20 x 16 in. (50.8 x 40.6cm) per compartment Model DI-1-2812 Bowl depth: 12 in. (30.5cm); Sink L x W: 28 x 20 in. (71.1 x 50.8cm)

Specifications

Specifications

Stainless Steel
1.5 in. IPS basket drain and aluminum/stainless mounting hardware to secure sink to countertop, deck mounted 8 in. swing spout faucet, 4 in. O.C
45 x 21 in. (114.3 x 53.3 cm)
114.3 cm
16 in.
40.64 cm
30.5 cm
19 kg
Drop-in Sink
2
45 in.
21 in.
53.3 cm
12 in.
42 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit