Learn More
Motic Swift Line Advanced Compound Microscopes Products for Science Education
Motic Swift Line AX500 Series Advanced Compound Microscopes feature LED illumination and an ergonomic professional frame with low-position coaxial focusing for a wide range of university and laboratory use.
$800.00 - $975.00
Specifications
For Use With (Application) Research, University, Laboratory, Advanced Biology
Magnification Power 40X, 100X, 400X, 1000X
Head Style Binocular
View More Specs
Products 3
Catalog Number Mfr. No. Objective Types Price Quantity & Availability  
Catalog Number Mfr. No. Objective Types Price Quantity & Availability  
S81365 Products for Science Education
View Documents
Motic Swift Line
AX502A
Achromatic
Each for $800.00
Add to cart
S81367 Products for Science Education
View Documents
Motic Swift Line
AX502PL
Plan
Each for $975.00
Add to cart
S81375 Products for Science Education
View Documents
Motic Swift Line
AX502ASC
Super High Contrast
Each for $900.00
Add to cart
Description
Description
  • Choice of achromat, super high contrast, or plan optics
  • Inward-facing quadruple revolving nosepiece
  • Variable 3W LED illumination
  • Built-in low-drive mechanical stage with extended post for X and Y control (135 x 140 mm)
  • 18 mm eyepieces
  • 30° inclined ergonomic Siedentopf viewing head
  • Built-in internal 110V to 220 V auto-switching power supply
Specifications
Specifications
Research, University, Laboratory, Advanced Biology
40X, 100X, 400X, 1000X
Binocular