Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Advanced Containment Systems Quick Flate Air Inflatable Emergency Shelters
Rugged ethylene propylene diene monomer (EPDM) Air Beam structure is lightweight, resilient, and stable
Manufacturer:  Advanced Containment Systems, Inc. (ACSI)Supplier Diversity Partner QF1015D
 View more versions of this product
Catalog No. 19-130-3907
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Two-line decontamination shower facilities including inlet and outlet connections for optional air and water systems
96 in.
180 in.
140 lb.
304.8cm
180 x 120 x 96 in. (457.2 x 304.8 x 243.84cm)
243.84cm
457.2cm
Decon System
63.5kg
120 in.