Learn More
Osmometer Calibration Standards
Calibration standards for use with Advanced osmometers
Manufacturer:  Advanced Instruments 3LA090
 View more versions of this product
Catalog No. 22-046710
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Osmometer calibration standard
Liquid
900mOsm/kg
3D3 osmometers
1 x 110mL
Clear
Sodium Chloride 0.15% to 9.5%, 2-Hydroxybiphenyl Sodium Salt Tetrahydrate <0.1%
Osmometer instrument calibration and quality control
Room Temperature