Osmometer Standards

For in vitro diagonostic use

Manufacturer: Advanced Instruments 3MA040

 View more versions of this product

Catalog No. 22-046-609

  Check Availability
Add to Cart

    Specifications

    Product Type Osmometer calibration standard
    Concentration 400mOsm/kg H2O
    Storage Requirements 2° to 30°C
    Quantity 10 x 2mL,10 x 2mL
    For Use With (Application) To check the performance of your osmometer and test methods, in-vitro diagnostic use
    Includes 2- Hydroxybiphenyl sodium salt tetrahydrate, 0.01% added as preservative
    For Use With (Equipment) 3250, 3900 and A2O Osmometers

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit