Learn More
Protinol™ Serum Osmometer Controls
Quality control check for the clinical laboratory for osmometers from any manufacturer
Manufacturer:  Advanced Instruments 200215
 View more versions of this product
Catalog No. 22-046-603
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
280mOsm Serum control kit
280mOsm/kg H2O
Serum osmometer
2° to 8°C
2mL Vials
Use in evaluating performance of osmometer in clinical laboratories testing of Human Serum samples
NIST, ISO 9000, CLIA, CAP
8 Pack
Safety and Handling
ShelfLife : Unopened: 18 months; Opened: 7 days when refrigerated