Learn More
Renol™ Urine Control Solution
Manufacturer:  Advanced Instruments 200217
 View more versions of this product
Catalog No. 22-046-605
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
2 Level Urine control kit
300 and 800mOsm/kg H2O
Urine osmometer
2° to 8°C
2mL Vials and 4 Vials per level
Evaluating the performance the osmometer in clinical laboratories testing Human Urine
CE, CLIA, CAP
8 Pack
Safety and Handling
ShelfLife : Unopened: 2 Years; Opened: 10 days when refrigerated