Renol™ Urine Control Solution

Manufacturer: Advanced Instruments 200219

 View more versions of this product

Catalog No. 22-046-607

  Check Availability
Add to Cart

    Specifications

    Product Type Urine control kit
    Concentration 800mOsm/kg H2O
    For Use With (Equipment) Urine osmometer
    Certifications/Compliance CE, CLIA, CAP
    Shelf Life Unopened: 2 Years; Opened: 10 days when refrigerated
    Storage Requirements 2° to 8°C
    Quantity 8 Pack,8 Pack
    For Use With (Application) Evaluating the performance the osmometer in clinical laboratories testing Human Urine
    Includes 2mL vials

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit