Learn More
Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Filter Screw Cap
For maintenance of adherent cells or suspension cell culture. Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Filter Screw Cap provides optimal conditions for cultivation of sensitive and fastidious cells.
$881.00 - $1397.00
Catalog Number | Mfr. No. | Culture Area | Volume (Metric) Working | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Culture Area | Volume (Metric) Working | Price | Quantity & Availability | ||||
07-000-666
|
Greiner Bio-One
661975 |
175cm2 | 650mL |
Case of 40 for $881.00
|
|
||||
07-000-655
|
Greiner Bio-One
658975 |
75cm2 | 250mL |
Case of 120 for $1,397.00
|
|
||||
07-000-171
|
Greiner Bio-One
660975 |
175 cm2 | 20 to 45 mL |
Case of 50 for $959.00
|
|
||||
Description
- Made of high-grade polystyrene
- Free of detectable DNases, RNases, human DNA, endotoxins and are non-cytotoxic
- Stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator
- Printed graduation for easier filling on both sides
- Provides homogeneous and optimized cell growth
Specifications
Polystyrene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.