Learn More
Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Standard Screw Cap
For maintenance of adherent cells or suspension cell culture. Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Filter Screw Cap provides optimal conditions for cultivation of sensitive and fastidious cells.
Manufacturer:  Greiner Bio-One 661960
 View more versions of this product
Catalog No. 07-000-665
Add to cart
Description
Description
  • Made of high-grade polystyrene
  • Free of detectable DNases, RNases, human DNA, endotoxins and are non-cytotoxic
  • Stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator
  • Printed graduation for easier filling on both sides
  • Provides homogeneous and optimized cell growth
Specifications
Specifications
Polystyrene
Standard screw cap
650mL
Advanced TC
175cm2