Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 4000 Full Facepiece Respirators, Push-To-Connect Adapter

Full-face reusable respirators with push-to-connect adapter are ideal for a variety of applications. Advantage™ 4000 series is compatible with several MSA respirator models, including air-purifying respirators, supplied-air respirators and powered air-purifying respirators.

$404.80 - $875.60

Specifications

Includes Push-to-connect Adapter
Suspension Type 4-point
Material (Lens) Polycarbonate
For Use With (Application) Utilities, mining, oil and gas, fire service, construction, confined space
Certifications (NIOSH) NIOSH 42 CFR Part 84
View More Specs

Products 12
Catalog Number Mfr. No. Size Material Harness Material Price Quantity & Availability  
Catalog Number Mfr. No. Size Material Harness Material Price Quantity & Availability  
18-139-401
View Documents
MSA™
10075915
Small Hycar™ Polyester Net Head Harness
Each for $404.80
Add to cart
 
18-139-403
View Documents
MSA™
10075911
Small Hycar™ Rubber
Each for $411.40
Add to cart
 
18-139-400
View Documents
MSA™
10075914
Small Silicone Polyester Net Head Harness
Each for $875.60
Add to cart
 
18-139-402
View Documents
MSA™
10075910
Small Silicone Rubber
Each for $411.40
Add to cart
 
18-139-405
View Documents
MSA™
10075909
Medium Hycar™ Polyester Net Head Harness
Each for $404.80
Add to cart
 
18-139-407
View Documents
MSA™
10075905
Medium Hycar™ Rubber
Each for $404.80
Add to cart
 
18-139-404
View Documents
MSA™
10075908
Medium Silicone Polyester Net Head Harness
Each for $462.55
Add to cart
 
18-139-406
View Documents
MSA™
10075904
Medium Silicone Rubber
Each for $411.40
Add to cart
 
18-139-409
View Documents
MSA™
10075921
Large Hycar™ Polyester Net Head Harness
Each for $404.80
Add to cart
 
18-139-411
View Documents
MSA™
10075917
Large Hycar™ Rubber
Each for $404.80
Add to cart
 
18-139-408
View Documents
MSA™
10075920
Large Silicone Polyester Net Head Harness
Each for $875.60
Add to cart
 
18-139-410
View Documents
MSA™
10075916
Large Silicone Rubber
Each for $411.40
Add to cart
 
Description

Description

  • Single-port facepiece with push-to-connect adapter
  • Compatible with MSA air-purifying respirators, supplied-air respirators and powered air-purifying respirators
  • NIOSH-approval 42 CFR Part 84
  • 4-pt suspension with rubber or polyester net head harness
  • Standand fog-reducing nosecup
  • Facepiece material is available in silicone or Hycar™
  • Three standard sizes to ensure a secure fit
  • Components lock into place in seconds for quick, convenient performance
  • Ideal for use in utilities, mining, oil and gas, fire service, construction, confined space, asbestos abatement, welding and sanding/grinding applications
Specifications

Specifications

Push-to-connect Adapter
Polycarbonate
NIOSH 42 CFR Part 84
4-point
Utilities, mining, oil and gas, fire service, construction, confined space
Full Facepiece Respirator with Adapter
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit