Description
The Advantage 900 Elastomeric half-mask respirator provides healthcare professionals, essential workers and first responders a secure protection solution with enhanced communication.
Elastomeric respirators are designed for repeated cleaning and decontamination, resulting in dramatic cost savings over filtering facepiece respirators. The included mechanical speaking diaphragm allows for improved communication among essential workers, patients and the public.
- No exhalation valve, allowing for filtration of exhaled air and providing a means of source control and use in a sterile field environment
- Reuse and cleaning capability make for a more eco-friendly alternative
- Overmolded MultiFlex nosecup provides stability on user's face while maintaining a secure fit
- Anthrocurve facepiece design maximizes fit and comfort and reduces potential leak points
Specifications
Specifications
| Connection Type | Bayonet |
| Color | Blue |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| Size | Large |
| With Exhalation Valve | No |
| Latex Free | Yes |
| Sterility | Non-sterile |