Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ohaus Corporation ADVENTURER™ PRECISION

Ready For Your Lab, Wherever That May Be.

Manufacturer:  Ohaus Corporation 30100631/RM

 View more versions of this product

Catalog No. 01-919-364


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. -With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.

Warranty and Services

2 Year

Specifications

Specifications

220g
NTEP
340mm
230mm
Cast Aluminum Base
5.8kg
50/60 Hz
Touchscreen LCD
RS232, USB
0.001g
AC Adapter
AX223N/E
2s
354mm
220g
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit