Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advillin Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209561AP

Catalog No. 50-173-3172


Add to cart

Description

Description

AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications

Specifications

Advillin
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75366, O88398
ADVIL, advillin, AVIL, DOC6
Rabbit
IgG
150 μL
-20°C
Primary
10677, 11567
Western Blot
Unconjugated
AVIL
Liquid
AVIL
Advillin Fusion Protein Ag15114
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit