Learn More
Azzota Corp AEE-788 DFS Item
Manufacturer:  Azzota Corp C1017
AEE-788 is an orally bioavailable multiple-receptor tyrosine kinase inhibitor. AEE788 inhibits phosphorylation of the tyrosine kinases of EGFR, HER2, and VEGF2. Please request if need more quantities
Catalog No. 50-161-3044