Learn More
Selleck Chemical LLCSupplier Diversity Partner AEE788 (NVP-AEE788) 50mg 497839-62-0  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S148650MG
AEE788 (NVP-AEE788) is a potent inhibitor of EGFR and HER2/ErbB2 with IC50 of 2 nM and 6 nM, less potent to VEGFR2/KDR, c-Abl, c-Src, and Flt-1, does not inhibit Ins-R, IGF-1R, PKCα and CDK1. Phase 1/2.
Catalog No. 50-677-6