Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AEG 3482, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
63735-71-7
C10H8N4O2S2
280.32
MQUYTXDAVCOCMX-UHFFFAOYSA-N
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
698112
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N
This item is not returnable. View return policy
63735-71-7
C10H8N4O2S2
280.32
MQUYTXDAVCOCMX-UHFFFAOYSA-N
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
698112
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N

Inhibitor of JNK signaling

Inhibitor of c-jun N-terminal kinase (JNK) signaling. Binds Hsp90 and facilitates HSF1 release, induces expression of Hsp70, which in turn blocks JNK activation and JNK-dependent apoptosis. Antiapoptotic; inhibits NGF withdrawal-induced death in SCG neurons (EC50 = 20 μM).
Quantity 10 mg
Formula Weight 280.33
Percent Purity >99%
Chemical Name or Material AEG 3482

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit