Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AEG 3482, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
63735-71-7
C10H8N4O2S2
280.32
MQUYTXDAVCOCMX-UHFFFAOYSA-N
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
698112
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N
This item is not returnable. View return policy
63735-71-7
C10H8N4O2S2
280.32
MQUYTXDAVCOCMX-UHFFFAOYSA-N
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
698112
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N

Inhibitor of JNK signaling

Inhibitor of c-jun N-terminal kinase (JNK) signaling. Binds Hsp90 and facilitates HSF1 release, induces expression of Hsp70, which in turn blocks JNK activation and JNK-dependent apoptosis. Antiapoptotic; inhibits NGF withdrawal-induced death in SCG neurons (EC50 = 20 μM).
Quantity 50 mg
Formula Weight 280.33
Percent Purity >99%
Chemical Name or Material AEG 3482

Recommended Storage : Store at Room Temperature