Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AEG 3482, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Inhibitor of c-jun N-terminal kinase (JNK) signaling. Binds Hsp90 and facilitates HSF1 release, induces expression of Hsp70, which in turn blocks JNK activation and JNK-dependent apoptosis. Antiapoptotic; inhibits NGF withdrawal-induced death in SCG neurons (EC50 = 20 μM).
Specifications
Quantity | 10 mg |
Formula Weight | 280.33 |
Percent Purity | >99% |
Chemical Name or Material | AEG 3482 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'