Tocris™ AEG 40730 dihydrochloride

IAP antagonist; induces apoptosis

Manufacturer: Tocris™ 5330/50

 View all options for this product

Catalog No. 53-305-0

  Check Availability
Add to Cart


For Research Use Only


    Description AEG 40730 dihydrochloride
    Molecular Weight 1120.06
    Purity >98
    Quantity 50mg
    Product Type AEG 40730 dihydrochloride

    IAP antagonist; binds to the BIR3 domain of cIAP1, cIAP2 and XIAP with nanomolar affinity. Reduces cIAP1, cIAP2 and XIAP protein levels in human breast cancer MDA-MB-231 cells. Induces apoptosis in combination with TNF, and potentiates TRAIL-mediated apoptosis in human colorectal carcinoma HCT 116 cells. Cell permeable.

    • Chemical Name: 222,Trifluoro-N-[[(2S)-1-(N-methyl-L-analyl-L-threonyl)-2-pyrrolidinyl]methyl]-N-(2-phenylethyl)acetamide, diethyl ether with 2,4-hexadiyne-1.6-diol dihydrochloride
    • Formula: C52H68F6N8O8.2HCl
    • Solubility: Soluble to 100mM in water and to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit