Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Aero™ Emergency Eye Wash Solution; Buffered pH Neutralizing, Stations and Bottles Products for Science Education

Bel-Art™ Aero™ Emergency Eye Wash Solution provides immediate first aid for the eyes. Effective in rinsing impurities and irritants from the eye and in preventing damage from acids and corrosive substances

Manufacturer:  Bel-Art™ F248825016/EMD

 View more versions of this product

Catalog No. S72484


Add to cart

Alert:

These products are available only in the United States.


Description

Description

Features:

  • Immediate response eye first aid for the laboratory, classroom, work site and public facilities
  • pH neutralizing solution helps the eye return to a normal 7.4pH level by quickly neutralizing corrosive chemicals
  • Instant access to fast, safe, and effective first aid for eye injuries caused by foreign particles such as dirt and debris, chemicals and other environmental irritants
  • All-in-one eye wash combines sterile saline with buffered, pH neutralizing solution to help relieve irritation, discomfort and minimize likelihood of permanent damage
  • 4 year shelf life from date of manufacture
  • Meets all FDA requirements for eye wash including buffered pH solution and preservative as per 21 CFR part 349
  • Unique eye-opener allows used fluids and contaminants to flow away from the eye (16oz and 32oz bottles)
  • Covered station keeps bottles clean throughout shelf life; carrying handle makes transport easy for field work; wall mountable, hardware included
  • Smaller 5oz. and 0.5oz. sizes can be easily included in portable work stations or in pockets for high risk situations
  • Quick twist top; easy-to-open and use
  • Meets ANSI Z358.1-2009 (as a personal eye wash only)
Specifications

Specifications

2 x 16 oz.
Clear
Unique Eye-Opener
Eyewash Station
473 mL
Rinsing impurities and irritants from the eye; Preventing damage from acids and corrosive substances
Yes
Safety and Handling

Safety and Handling

ShelfLife : 4 Years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit