Learn More
Axygen™ Aerosol Filter Tips, Sterile, Wide Bore
GSA_VA
Catalog No. 14222768
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 to 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance USP 3127
Clean Claims DNase, RNase, and Pyrogen Free
Format Rack
Tip Style Genomic/Wide Orifice
Packaging 100 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.