Learn More
Axygen™ Aerosol Filter Tips, Sterile, Wide Bore

Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
$964.00 - $1037.00
Specifications
Clean Claims | DNase, RNase, and Pyrogen Free |
---|---|
Material | PP (Polypropylene) |
Filtered | Filtered |
Color | Clear |
Sterility | Sterile |
Catalog Number | Mfr. No. | Volume (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Format | Price | Quantity & Availability | ||||
14-222-768
![]() |
Axygen™
TF1005WBRS |
100 to 1000 μL | Rack |
Case of 5000 for $1,037.00
|
|
14-222-780
![]() |
Axygen™
TF205WBRS |
1 to 200 μL | Rack |
Case of 4800 for $964.00
|
|
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
DNase, RNase, and Pyrogen Free | |
Filtered | |
Sterile |
PP (Polypropylene) | |
Clear | |
Genomic/Wide Orifice |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.