Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aerosol™ OT Solution, 75% (w/w), Fisher Chemical
Contains: Sodium Dioctyl Sulfosuccinate, Ethyl Alcohol, Water
Description
- Anionic surfactant solution of 75% sodium dioctyl sulfosuccinate in ethanol and water
- Efficient wetting agent, dispersant, and emulsifier for a variety of applications
- Used to reduce surface tension and to incrase absorbency and penetration
- Fisher Chemical also offers the 100% waxy solid surfactant (Catalog No.: A349)
Specifications
Aerosol™ OT Solution | |
75% (w/w) | |
78°C | |
72 to 78% | |
500 mL | |
Pass Test |
Undesignated | |
Liquid | |
1.09g/cm³ | |
5 to 7 | |
Dioctyl sulfosuccinate, AOT, DOSS | |
Glass Bottle |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction