Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AF 64394

Potent and selective GPR3 inverse agonist

$131.30 - $551.50

Specifications

CAS 1637300-25-4
Chemical Name or Material N-[[4-Chloro-2-(1-methylethoxy)phenyl]methyl]-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Molecular Formula C21H20ClN5O
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
69-311-0
View Documents
Tocris Bioscience™
6931/10
10 mg
Each for $131.30
Add to cart
 
69-315-0
View Documents
Tocris Bioscience™
6931/50
50 mg
Each for $551.50
Add to cart
 
Description

Description

  • Potent and selective GPR3 inverse agonist (pIC50 = 7.3)
  • Selective for GPR3 over GPR6 and GPR12 (pIC50 values are 5.1 and 4.9, respectively)
Specifications

Specifications

1637300-25-4
N-[[4-Chloro-2-(1-methylethoxy)phenyl]methyl]-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
C21H20ClN5O
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit