Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP Mouse anti-Human, Alexa Fluor™ 750, Clone: 16F50, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222004AF750

Catalog No. NP22204A750


Add to cart

Description

Description

AFAP Monoclonal antibody specifically detects AFAP in Human samples. It is validated for Flow Cytometry,ELISA.
Specifications

Specifications

AFAP
16F50
Flow Cytometry, ELISA
actin filament associated protein 1, FLJ56849
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
AFAP1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant aFAP. Mouse anti human aFAP Q12884.
0.1 mL
Primary
60312
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit