Learn More
AFAP1 Mouse anti-Human, Clone: OTI2G9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802329
Catalog No. 50-166-5944
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
AFAP1
Monoclonal
Unconjugated
AFAP1
Lyophilized
AFAP1
Human recombit protein fragment corresponding to amino acids 100-386 of human AFAP1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2G9
PBS with 8% trehalose and no preservative; pH 7.3
Q8N556
AFAP, AFAP-110, AFAP110
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
60312