Learn More
Selleck Chemical LLC
Afatinib (BIBW2992) 10mg 850140-72-6
Manufacturer: Selleck Chemical LLC S101110MG
Afatinib (BIBW2992) inhibits EGFR/ErbB irreversibly in vitro with IC50 of 0.5, 0.4, 10, 14, 1 nM for EGFRwt, EGFR L858R , EGFR L858R/T790M ErbB2 (HER2) and ErbB4 (HER4), respectively. Afatinib induces autophagy.