Learn More
Aat BioquestSupplier Diversity Partner AFC [7-Amino-4-trifluoromethylcoumarin] *CAS 53518-15-3* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 58
This compound is the precursor for a variety of AFC enzyme substrates. It is also used as a calibration standard for AFC-based enzyme substrates.
Catalog No. 50-195-2686