Learn More
Abnova™ AFG3L2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00010939T01
Catalog No. 89-932-795
Add to cart
Description
Description
Specifications
Specifications
293T
BC065016.1
pCMV-AFG3L2 full-length
100 μL
Human
AFG3L2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 87.78kDa
Transient overexpression cell lysate was tested with anti-AFG3L2 antibody (Mfr. No. H00010939-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot