Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFG3L2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271647AF647

Catalog No. N271647A647


Add to cart

Description

Description

AFG3L2 Monoclonal antibody specifically detects AFG3L2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AFG3L2
OTI4B6
Western Blot
Affinity Purified, Conjugated
AFG3L2
Recombinant protein fragment corresponding to amino acids 1-250 of human AFG3L2 (NP_006787) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
AFG3 (ATPase family gene 3, yeast)-like 2, AFG3 ATPase family gene 3-like 2 (S. cerevisiae), AFG3 ATPase family gene 3-like 2 (yeast), AFG3-like protein 2, ATPase family gene 3, yeast, EC 3.4.24, EC 3.4.24.-, FLJ25993, Paraplegin-like protein, SCA28, spinocerebellar ataxia 28
Mouse
IgG1
0.1 mL
Primary
10939.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit