Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aflatoxin Antibody (ATB), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108086
Description
Aflatoxin Monoclonal antibody specifically detects Aflatoxin in Nonspecies specific samples. It is validated for ELISA.Specifications
Aflatoxin | |
Monoclonal | |
Unconjugated | |
AFB1-AR1, Aflatoxin M1, AFT, Aldoketoreductase 7, FB1 | |
Aflatoxin | |
0.2 mg | |
Virology Bacteria and Parasites | |
This recognizes free aflatoxins B1 and B2.No cross-reactivity with G1, G2 and M1. | |
Store at 4C. Do not freeze. | |
IgG1 |
ELISA | |
ATB | |
ELISA 1:100-1:2000 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction