Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ AFMID Protein 2

Human Recombinant Protein

Manufacturer:  Novus Biologicals™ NBP194069PEP

Catalog No. NBP194069PE

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Amino Acid Sequence: LPFFLFFHGGYWQSGSKDESAFMVHPLTAQGVAVVIVAYGIAPKGTLDHMVDQVTRSVAFVQKRY
  • Source: E.Coli

This is a blocking peptide for NBP1-94069. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications

Specifications

0.5mg/mL
PBS and 1M Urea, pH 7.4.
25kDa
>80%
E.Coli
Store at -20°C. Avoid freeze-thaw cycles.
AFMID
Unlabeled
Yes
Blocking/Neutralizing, Control
125061
Chromatography
0.1mL
LPFFLFFHGGYWQSGSKDESAFMVHPLTAQGVAVVIVAYGIAPKGTLDHMVDQVTRSVAFVQKRY
RUO
AFMID
Human
Safety and Handling

Safety and Handling

ShelfLife : 365

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only