Learn More
Abnova™ AFP HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L006T6
Catalog No. 89-932-805
Add to cart
Description
Description
Specifications
Specifications
HEK293
BC027881
Theoretical M.W. 69kDa
Transient overexpression cell lysate was tested with anti-AFP antibody (Mfr. No. H00000174-M01) by Western Blot.
In modified RIPA Lysis Buffer.
2 mg/mL
AFP HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-AFP full length
100 μg
Human
Immunoprecipitation, Western Blot